missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules

Product Code. 10731841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10731841 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10731841 Supplier Thermo Scientific Acros Supplier No. 178780250
  • ONLINE EXCLUSIVE PRICE
    £93.70
    £84.65 / 25g
    Save £9.05
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9011-14-7
Physical Form Beads
Melting Point >150.0°C
Molecular Formula (C5H8O2)n
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
MDL Number MFCD00134349
Density 1.18
Synonym Methyl methacrylate, polymerized, PMMA
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 25 g
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
SMILES COC(=O)C(C)(-*)C-*
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
Molecular Weight (g/mol) NaN
Flash Point 303°C
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Particle Size Bead size (typically): 50 to 150 micron
Chemical Name or Material Poly(methyl methacrylate), M.W. 93300
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.