missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), approx. M.W. 15000 (GPC)

Product Code. 10541641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541641 50 g Plastic bottle 50g
10164400 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10541641 Supplier Thermo Scientific Acros Supplier No. 190690500
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
Synonym Methyl methacrylate, polymerized, PMMA
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Color White
Physical Form Beads
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Density 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 50 g
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Plastic bottle
Particle Size Bead size (typically): 10 to 100 micron
Chemical Name or Material Poly(methyl methacrylate), Average M.W. 15000 (GPC)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.