missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), beads, average M.W. 35,000

Product Code. 10711651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711651 500 g Glass bottle 500g
10701651 25 g Glass bottle 25g
10791641 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711651 Supplier Thermo Scientific Acros Supplier No. 178765000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
Synonym Methyl methacrylate, polymerized, PMMA
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Color White
Physical Form Beads
Melting Point 105.0°C
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Density 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 500 g
Flash Point 300°C
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Particle Size Bead size (typically): 50 to 150 micron
Refractive Index 1.49
Chemical Name or Material Poly(methyl methacrylate), nominal M.W. 38000
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.