missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), 600 micron

Product Code. 11355018
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355018 250 g 250g
11345018 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355018 Supplier Thermo Scientific Alfa Aesar Supplier No. 043969.30
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

Poly(methyl methacrylate), 600 microns is used in many medical and biochemical applications. It is suitable for 2D and 3D particle assemblies and for investigations in the colloidal crystal field due to the high monodispersity. It is also used in the field of one- and two-component toners for copying machines. Further, it is having a reduced non-specific protein binding activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Physical Form Powder
Solubility Information Soluble in tetrahydrofuran,toluene,cyclohexanone,ethyl acetate and chloroform. Insoluble in water,alcohols and aliphatic hydrocarbons.
Quantity 250 g
Particle Size 600 Micron
Chemical Name or Material Poly (methyl methacrylate)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.