missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(methyl hydrogen siloxane), TRC
Specifications
Specifications
| Chemical Name or Material | Poly(methyl hydrogen siloxane) |
| CAS | 63148-57-2 |
| Molecular Formula | (CH 3 ) 3 SiO[(CH 3 )HSiO] n Si(CH 3 ) 3 |
| Quantity | 2.5 g |
| Synonym | Methyl Hydrogen Silicones, Methyl Hydrogen Siloxanes, Methyl Hydrogen Trimethylsilyl-terminated Polysiloxanes, Methyl Hydrogen Polysiloxane, Momentive SS 4300 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?