missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl hydrogen siloxane), TRC

Product Code. 30422298
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422298 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422298 Supplier Toronto Research Chemicals Supplier No. TRCP6884655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Poly(methyl hydrogen siloxane)
CAS 63148-57-2
Molecular Formula (CH 3 ) 3 SiO[(CH 3 )HSiO] n Si(CH 3 ) 3
Quantity 5 g
Synonym Methyl Hydrogen Silicones, Methyl Hydrogen Siloxanes, Methyl Hydrogen Trimethylsilyl-terminated Polysiloxanes, Methyl Hydrogen Polysiloxane, Momentive SS 4300
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.