missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400

Product Code. 11329308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329308 5 g 5g
11339308 25 g 25g
11319308 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329308 Supplier Thermo Scientific Alfa Aesar Supplier No. 043228.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fluorosilicone lubricant | CAS: 63148-56-1 | C4H7F3OSi | 156.179 g/mol

Fluorosilicone lubricantPoly(methyl-3,3,3-trifluoropropylsiloxane) is used in the preparation of siliconized- glass fiber sheets, which acts as a support for protein-chemical analysis of electroblotted proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-56-1
Molecular Formula C4H7F3OSi
Molecular Weight (g/mol) 156.179
MDL Number MFCD00241477
InChI Key CUMCLKHSZZGALD-UHFFFAOYSA-N
Synonym methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra
PubChem CID 4285173
IUPAC Name methyl-oxo-(3,3,3-trifluoropropyl)silane
SMILES C[Si](=O)CCC(F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Density 1.25 g/mL
Solubility Information Immiscible with water.
Quantity 5 g
Flash Point 260°C (500°F)
Refractive Index 1.381
Chemical Name or Material Poly (methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.