missing translation for 'onlineSavingsMsg'
Learn More

Poly-L-Lysine Hydrobromide [MW 1,500-8,000], MP Biomedicals

Product Code. 17328583 Shop All MP Biomedicals Products
Change view
Click to view available options
:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17328583 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17328583 Supplier MP Biomedicals Supplier No. 0215268980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 25988-63-0
Molecular Formula C18H38N6O4
Synonym lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
Quantity 100 mg
InChI Key WBSCNDJQPKSPII-KKUMJFAQSA-N
SMILES C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N
IUPAC Name (2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
Molecular Weight (g/mol) 402.54
PubChem CID 72363
Formula Weight 1,500 to 8,000
Chemical Name or Material Poly-L-Lysine Hydrobromide [MW 1, 500-8, 000]
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.