missing translation for 'onlineSavingsMsg'
Learn More

Poly(I:C) (~90%), TRC

Product Code. 30423843
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423843 1 g -
30410498 10 mg 10mg
30410896 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423843 Supplier Toronto Research Chemicals Supplier No. TRCP6880751G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24939-03-5
Molecular Formula H2(C10 H11 N4 O8 P)n . H2(C9 H12 N3 O8 P)n
Molecular Weight (g/mol) 671.4
Synonym 5'-Inosinic Acid Homopolymer Complex with 5'-Cytidylic Acid Homopolymer, 5'-Inosinic Acid Polymers Complex with 5'-Cytidylic Acid Polymers, 5'-Cytidylic Acid Homopolymer Complex with 5'-Inosinic Acid Homopolymer, 5'-Cytidylic Acid Polymers Complex with 5'-Inosinic Acid Polymers, 5'-Inosinic Acid Homopolymer-5'-cytidylic Acid Homopolymer Complex, NSC 120949, NSC 123233, NSC 128576, Poly(C).poly(I), Poly(C:I), Poly(I)-poly(C), Poly(I).poly(C), Poly(I).poly(C) Complex, Poly(cytidylic acid)-poly(inosinic acid), Poly(rI)-poly(rC), Poly(rI).poly(rC), Poly(rI):poly(rC), Poly(rI:rC), Polycytidylic-polyinosinic Acid, Polyinosinate-polycytidylate Complex, Polyinosinate:polycytidylate, Polyinosine-polycytidylic Acid, Polyinosinic Acid:polycytidylic Acid, Polyinosinic Acid:polycytidylic Acid Complex, Polyinosinic-polycytidylic Acid, Polyinosinic-polycytidylic Acid Complex, Polyribocytidylic Acid-polyriboinosinic Acid Complex
SMILES O[C@@H]1[C@H](O)[C@@H](COP(O)(O)=O)O[C@H]1N2C(NC=NC3=O)=C3N=C2.O[C@@H]4[C@H](O)[C@@H](COP(O)(O)=O)O[C@H]4N5C=CC(N)=NC5=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H13N4O8P.C9H14N3O8P/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)
Formula Weight 671.099
Chemical Name or Material Poly(I:C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.