missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Poly(I:C)

Product Code. 30001395
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001395 10 mg 10mg
18035712 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001395 Supplier R&D Systems™ Supplier No. 4287/10
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. TLR3 agonist

Poly(I:C) is a synthetic double-stranded RNA (dsRNA); endosomal Toll-like receptor 3 (TLR3) agonist. It also binds to the cytoplasmic receptors, retinoic acid-inducible gene I (RIG-I) and melanoma differentiation-associated gene 5 (MDA-5), elicits the secretion of type I interferon (IFN) and pro-inflammatory cytokines. Poly(I:C) is commonly used for immune system activation in vivo. Poly(I:C) also promotes maturation of hPSC-derived cardiomyocytes by modulating early Notch signaling and cardiomyogenic transcriptional programs. Poly(I:C) promotes generation of stable, mature dendritic cells in combination with TLR8 agonist CL 075 . Transfection into NIT-1 β cells induce β-cell apoptosis. Commonly used to stimulate immune system activation in vivo. Also promotes maturation of hPSC-derived cardiomyocytes.

To prevent denaturation, this product should be reconstituted in solutions with physiological salt concentrations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24939-03-5
Molecular Formula C19H27N7O16P2

Specifications

Quantity 10 mg
Product Line Tocris
Purity >98%
Molecular Weight (g/mol) 671.406
Chemical Name or Material Poly(I:C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.