missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), approx. M.W. 100,000, ACROS Organics™

Brand:  Acros Organics 178580050

Code : 29

Additional Details : CAS Number : 25322-68-3 Weight : 0.06000kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10062290

  • £18.07 / 5g


In stock, ready to ship: 16
Add to basket

Description

Description

Specifications

Chemical Identifiers

25322-68-3
MFCD00081839
PEO

Specifications

Poly(ethylene oxide)
25322-68-3
MFCD00081839
PEO
Powder
Glass bottle
Solubility in water: soluble. Other solubilities: soluble in many organic solvents
65.0°C to 67.0°C
Approximate M.W. 100.000
(-CH2CH2O-)n
15, 7688
Authentic
1% max.
1.4539
White to Yellow
5g
Videos
SDS Documents
Certificates