missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol methyl ether), average M.W. 750

Product Code. 10164210 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
500g
5g
This item is not returnable. View return policy

Product Code. 10164210

Brand: Thermo Scientific Acros 192320010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
SMILES COCCO

Specifications

Color White
pH 4 to 7 (5% aq. soln.)
Physical Form Fused Mass
Avg. Mol. Wt. or Mol. Wt. Range 715 to 785
Linear Formula CH3(OCH2CH)nOH
Density 1.09g/mL
Quantity 1 kg
IUPAC Name 2-methoxyethanol
Hydroxyl Value 71.5 to 78.5mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.1% max.
Chemical Name or Material Poly(ethylene glycol methyleether), Average M.W. 750
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.