missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 6000

Product Code. p-4599881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
10g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10154210 10 g Glass bottle 10g
10523561 1 kg Plastic bottle 1kg
10751604 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10154210 Supplier Thermo Scientific Acros Supplier No. 192280100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form White flakes or powder
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 10 g
Formula Weight 6000
Hydroxyl Value 17 to 20mg KOH/g
Packaging Glass bottle
Chemical Name or Material Poly(ethylene glycol)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.