missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 4000

Product Code. p-4229325 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic drum
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10783902 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10783902 Supplier Thermo Scientific Acros Supplier No. 434630050
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Flakes
Melting Point 54.0°C to 58.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15,7688
Quantity 5 kg
Formula Weight 4000
Infrared Spectrum Authentic
Packaging Plastic drum
Water 0.5% max.
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.