missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 300

Product Code. 10797982 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
This item is not returnable. View return policy

Product Code. 10797982

Brand: Thermo Scientific Acros 192220025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color Clear liquid
Melting Point -15.0°C to +10.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1200g/mL
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 2.5 L
Flash Point >180°C
Hydroxyl Value 356 to 394mg KOH/g
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Specific Gravity 1.12
Viscosity 85-100 mPa.s (20°C)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.