missing translation for 'onlineSavingsMsg'
Learn More

Poly(Ethylene Adipate)~ 10,000 By GPC, TRC

Product Code. 30380084
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380084 500 mg -
30379099 100 mg 100mg
30382174 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380084 Supplier Toronto Research Chemicals Supplier No. TRCE249380500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24938-37-2
Molecular Formula C6H10O4 . C2H6O2
Synonym 1,2-Ethanediol-adipic Acid Copolymer, Adipic Acid-1,2-ethanediol Copolymer, Adipic Acid-ethylene Glycol Copolymer, Adipic Acid-ethylene Glycol Polyester, Adipic Acid-ethylene Glycol Polymer, Hexanedioic acid-ethylene glycol copolymer, PEGA, Poly(Ethylene Glycol Adipate), Ethylene Glycol-adipic Acid Polymer
IUPAC Name bis(2-hydroxyethyl) hexanedioate
SMILES OCCOC(=O)CCCCC(=O)OCCO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H18O6/c11-5-7-15-9(13)3-1-2-4-10(14)16-8-6-12/h11-12H,1-8H2
Formula Weight 234.1103
Chemical Name or Material Poly(Ethylene Adipate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.