missing translation for 'onlineSavingsMsg'
Learn More

Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da

Product Code. 11340290 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11340290

Brand: Thermo Scientific Acros 436200050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 26780-50-7 | (C3H4O2)x(C2H2O2)y | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26780-50-7
Molecular Formula (C3H4O2)x(C2H2O2)y
Molecular Weight (g/mol) NaN
MDL Number MFCD00131930
InChI Key LCSKNASZPVZHEG-UHFFFAOYNA-N
Synonym vicryl, poly d,l-lactide-co-glycolide, polyglactin 370 usan, polyglactin, 1,4-dioxane-2,5-dione-3,6-dimethyl-1,4-dioxane-2,5-dione 1:1, polyglactin 910 usan, vicryl rapide, vicryl plus
PubChem CID 71391
IUPAC Name 1,4-dioxane-2,5-dione; 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC(-*)C(=O)OCC(=O)O-*

Specifications

Color Tan to White
Physical Form Crystalline Powder or Granules
Solubility Information Solubility in water: soluble.
Quantity 5 g
Packaging Glass bottle
Viscosity 0.15 to 0.25 dL/g
Chemical Name or Material Poly(DL-lactide-co-glycolide)
Is Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da acid-terminated or ester-terminated?

Our Poly(DL-lactide-co-glycolide), 50:50 lactide glycolide, M.N. 5,000 to 10,000 Da is acid-terminated.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.