missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-D-Lysine Hydrobromide, MP Biomedicals
£56.87 - £1220.00
Chemical Identifiers
CAS | 27964-99-4 |
---|---|
Molecular Formula | C6H15BrN2O2 |
Molecular Weight (g/mol) | 227.102 |
InChI Key | MEXAGTSTSPYCEP-NUBCRITNSA-N |
Synonym | d-lysine hydrobromide |
PubChem CID | 87493163 |
IUPAC Name | (2R)-2,6-diaminohexanoic acid;hydrobromide |
SMILES | C(CCN)CC(C(=O)O)N.Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428350
|
MP Biomedicals
0210269410 |
10 mg |
£56.87
10mg |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
17318583
|
MP Biomedicals
0210269450 |
50 mg |
£142.04
50mg |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||||
11448350
|
MP Biomedicals
0210269480 |
100 mg |
£296.80
100mg |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
11458350
|
MP Biomedicals
0210269490 |
500 mg |
£643.42
500mg |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
11468350
|
MP Biomedicals
0210269491 |
1 g |
£1220.00
1g |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
Description
Chemical Identifiers
27964-99-4 | |
227.102 | |
d-lysine hydrobromide | |
(2R)-2,6-diaminohexanoic acid;hydrobromide |
C6H15BrN2O2 | |
MEXAGTSTSPYCEP-NUBCRITNSA-N | |
87493163 | |
C(CCN)CC(C(=O)O)N.Br |
Specifications
27964-99-4 | |
d-lysine hydrobromide | |
C(CCN)CC(C(=O)O)N.Br | |
227.102 | |
4,000 to 15,000 |
C6H15BrN2O2 | |
MEXAGTSTSPYCEP-NUBCRITNSA-N | |
(2R)-2,6-diaminohexanoic acid;hydrobromide | |
87493163 | |
Poly-D-Lysine Hydrobromide |
Safety and Handling
Recommended Storage : 0°C