missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Poly Crimp™ Seals

Product Code. 10626991
Click to view available options
Material (Liner):
PTFE
PTFE/Silicone with Slit
PTFE/Silicone with Starburst
PTFE/Silicone/PTFE
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 10626991

Brand: JG Finneran Associates™ 551011

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Material (Liner) PTFE
Color Clear
Material PTFE
Quantity 1000/Pk.
Thickness (Metric) 10 mm
Closure Size 11 mm
For Use With (Equipment) 12 x 32 mm Sample Vials with Crimp Style Finish
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.