missing translation for 'onlineSavingsMsg'
Learn More

Poly(caprolactone) triol, average M.W. 300, Thermo Scientific Chemicals

Catalog Number 10781081
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
39280-09-6
MFCD00084405
This item is not returnable. View return policy
39280-09-6
MFCD00084405

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Viscous Liquid
Density 1.0700g/mL
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass Bottle
Refractive Index 1.4750 to 1.4780 (20°C, 589nm)
Chemical Name or Material Poly(caprolactone) triol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'