missing translation for 'onlineSavingsMsg'
Learn More

Poly(caprolactone) diol, average M.N. 2000

Product Code. 11966981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11966981 2.5 kg Glass Bottle 2.5kg
10245570 250 g Glass Bottle 250g
10503171 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11966981 Supplier Thermo Scientific Acros Supplier No. 183210025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36890-68-3 | (C6H10O2)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36890-68-3
Molecular Formula (C6H10O2)n
MDL Number MFCD23713083
SMILES *-CCCCCC(=O)O-*

Specifications

Color White
Physical Form Waxy Solid
Density 1.0700g/mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in thf
Quantity 2.5 kg
Infrared Spectrum Authentic
Loss on Drying 2% max.
Packaging Glass Bottle
Specific Gravity 1.07
Chemical Name or Material Poly(caprolactone) diol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.