missing translation for 'onlineSavingsMsg'
Learn More

Poly(Allylamine Hydrochloride)

Product Code. 11374748
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11374748

Brand: Thermo Scientific Alfa Aesar 043092.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

As a polymeric primary amine; Forms hydrogels | CAS: 71550-12-4 | C3H7N | 93.56 g/mol

Poly(allylamine hydrochloride) is used in film and polymer industries and cell encapsulation. Further, it is used to make redox hydrogel- modified electrodes for measuring enzyme responses. In addition, it is used in the preparation of an antireflective coating consists of alternate layers of poly(allyamine hydrochloride) and silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71550-12-4
Molecular Formula C3H7N
Molecular Weight (g/mol) 93.56
MDL Number MFCD00054329
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine hydrochloride, 3-aminopropene hydrochloride, poly allylamine hydrochloride, allylamine, hydrochloride, allylammonium chloride, 2-propen-1-amine, hydrochloride, unii-r2h25ilf98, 2-propen-1-amine hydrochloride, allylaminehydrochloride, prop-2-en-1-amine hydrochloride
PubChem CID 82291
IUPAC Name prop-2-en-1-amine;hydrochloride
SMILES NCC=C

Specifications

Color Yellow
Physical Form Powder
Solubility Information Soluble in water.
Quantity 50 g
Chemical Name or Material Poly (Allylamine Hydrochloride)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.