missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 25 wt.% solution in water; ∽ M.W. 240,000, ACROS Organics™

Brand:  Acros Organics 185012500

Code : 29

Additional Details : CAS Number : 9003-01-4 Weight : 0.30000kg

Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10317510

  • £64.60 / 250g


EU Stock 11
Add to basket

Specifications

Chemical Identifiers

Poly(acrylic acid)
7732-18-5
[-CH2CH(CO2H)-]n
25 wt. % Solution in Water
23 to 27% (solids)
MFCD00084394

Specifications

Poly(acrylic acid)
7732-18-5
Liquid
[-CH2CH(CO2H)-]n
Authentic
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
100.0°C
250g
25 wt. % Solution in Water
1.0900g/mL
23 to 27% (solids)
MFCD00084394
Glass bottle
1.09
Clear
Videos
SDS