missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000

Product Code. 10174430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174430 500 g Glass bottle 500g
10357740 5 g Glass bottle 5g
10675432 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10174430 Supplier Thermo Scientific Acros Supplier No. 178045000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9003-05-8 | (C3H5NO)n | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-05-8
Molecular Formula (C3H5NO)n
MDL Number MFCD00084392
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C(-*)C-*

Specifications

Color White to Yellow
Physical Form Granules
Linear Formula [-CH2CH(CONH2)-]n
Density 1.3000g/mL
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethylene glycol, glycerol, lactic acid
Quantity 500 g
IUPAC Name prop-2-enoic acid
Molecular Weight (g/mol) 72.06
PubChem CID 6581
ChEBI CHEBI:18308
Formula Weight 5 to 6.000.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 13% max.
Packaging Glass bottle
Specific Gravity 1.3
Chemical Name or Material Poly(acrylamide)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.