missing translation for 'onlineSavingsMsg'
Learn More

Poly(4-vinylpyridine), cross-linked, TRC

Product Code. 30417963
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417963 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417963 Supplier Toronto Research Chemicals Supplier No. TRCP6893805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Poly(4-vinylpyridine)
CAS 9017-40-7
Quantity 5 g
Synonym 4-Vinyl-pyridine polymer with divinylbenzene, Diethenyl-benzene polymer with 4-ethenylpyridine, 4-Vinylpyridine-divinylbenzene copolymer, CR 2, CR 2 (vinyl polymer), Divinylbenzene-4-ethenylpyridine copolymer, Divinylbenzene-4-vinylpyridine copolymer, Divinylbenzene-4-vinylpyridine polymer, Reillex 402, Reillex 425, Reillex X 425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.