missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(4-vinylpyridine), cross-linked, TRC
Specifications
Specifications
| Chemical Name or Material | Poly(4-vinylpyridine) |
| CAS | 9017-40-7 |
| Quantity | 10 g |
| Synonym | 4-Vinyl-pyridine polymer with divinylbenzene, Diethenyl-benzene polymer with 4-ethenylpyridine, 4-Vinylpyridine-divinylbenzene copolymer, CR 2, CR 2 (vinyl polymer), Divinylbenzene-4-ethenylpyridine copolymer, Divinylbenzene-4-vinylpyridine copolymer, Divinylbenzene-4-vinylpyridine polymer, Reillex 402, Reillex 425, Reillex X 425 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?