missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(2-ethyl-2-oxazoline), M.W. 500,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Off-white solid |
Solubility Information | Soluble in water and organic solvents. |
Quantity | 100 g |
Chemical Name or Material | Poly (2-ethyl-2-oxazoline), M.W. 500,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'