missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-ethyl-2-oxazoline), M.W. 50,000

Product Code. 11354538
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354538 100 g 100g
11344538 500 g 500g
11334538 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354538 Supplier Thermo Scientific Alfa Aesar Supplier No. 042396.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol

Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25805-17-8
Molecular Formula (C5H9NO)n
Molecular Weight (g/mol) 99.13
MDL Number MFCD00134377
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC(=O)N(-*)CC-*

Specifications

Color White
Physical Form Off-white solid
Solubility Information Soluble in water and organic solvents.
Quantity 100 g
Chemical Name or Material Poly (2-ethyl-2-oxazoline), M.W. 50,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.