missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-ethyl-2-oxazoline), M.W. 200,000

Product Code. 11384538
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11384538

Brand: Thermo Scientific Alfa Aesar 042397.A4

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol

Poly(2-ethyl-2-oxazoline) shows the application in biocompatibility, avoiding accumulation in tissue, and rapid clearance from the bloodstream. It is also adapted to many different functions for both protein and small drug conjugation. It also acts as a perfect substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). It is also used as an adhesion promoter in coatings and heat sealing. It behaves as a hydrophilic block and utilized in the synthesis of amphiphilic diblock copolymers by reacting with poly(1,3-trimethylene carbonate) (PTMC) as a hydrophobic block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25805-17-8
Molecular Formula (C5H9NO)n
Molecular Weight (g/mol) 99.13
MDL Number MFCD00134377
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC(=O)N(-*)CC-*

Specifications

Physical Form Off-white solid
Solubility Information Soluble in water and organic solvents.
Quantity 2.5 kg
Chemical Name or Material Poly (2-ethyl-2-oxazoline), M.W. 200,000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.