missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR3K Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552220
This item is not returnable.
View return policy
Description
Immunogen sequence: PGCGNGLIVE EGQRCHRFAC NTCPYVHNIT RKVTNRKYPK LKEVDDVLGG AAAWENVDST AESCPKCEHP RAYFMQLQTR SADEPMTTFY KCCNAQCGHR WRD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Eukaryotes produce 3 distinct classes of RNA polymerase, Pol I, II and III. Each polymerase is responsible for the synthesis of a different class of RNA. RNA polymerase III (Pol III) transcribes the 5S rRNA genes and all of the tRNA (transfer RNA) genes. POLR3K, also known as DNA-directed RNA polymerase III subunit RPC11, is a 108 amino acid nuclear protein that is one of the seventeen subunits that comprise Pol III. Specifically, POLR3K is a DNA-dependent RNA polymerase that uses the four ribonucleoside triphosphates to catalyze the transcription of DNA to RNA. The gene encoding POLR3K maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs and is associated with a variety of genetic disorders.
Specifications
| POLR3K | |
| Polyclonal | |
| Unconjugated | |
| POLR3K | |
| 12.3kDa; 1500004O14Rik; AI196849; AU014893; C11; C11-RNP3; DNA-directed RNA polymerase III subunit K; DNA-directed RNA polymerase III subunit RPC10; DNA-directed RNA polymerases III 12.5 kDa polypeptide; estrous-specific protein, 250 kDa; hRPC11; HsC11p; My010; POLR3K; polymerase (RNA) III (DNA directed) polypeptide K; polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa; polymerase (RNA) III subunit K; RNA polymerase III 12.5 kDa subunit; RNA polymerase III subunit C10; RNA polymerase III subunit C11; RNA polymerase III subunit CII; RNA polymerase III subunit K; RPC10; RPC11; RPC12.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51728 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2Y1 | |
| POLR3K | |
| Recombinant protein corresponding to Human POLR3K. Recombinant protein control fragment (Product #RP-89380). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction