missing translation for 'onlineSavingsMsg'
Learn More

POLR3K Antibody [DyLight 350], Novus Biologicals™

Product Code. 30762293 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762293 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762293 Supplier Novus Biologicals Supplier No. NBP346938UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POLR3K Polyclonal antibody specifically detects POLR3K in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3K
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C11, DNA-directed RNA polymerase III subunit K, DNA-directed RNA polymerase III subunit RPC10, DNA-directed RNA polymerases III 12.5 kDa polypeptide, hRPC11, HsC11p, polymerase (RNA) III (DNA directed) polypeptide K (12.3 kDa), polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa, RNA polymerase III 12.5 kDa subunit, RNA polymerase III subunit (hRPC11), RNA polymerase III subunit C10, RNA polymerase III subunit C11, RNA polymerase III subunit CII, RPC10, RPC11C11-RNP3, RPC12.5
Host Species Rabbit
Immunogen polymerase(RNA) III(DNA directed) polypeptide K, 12.3 kDa. (Uniprot# Q9Y2Y1)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51728
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.