missing translation for 'onlineSavingsMsg'
Learn More

POLR3K Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-94027-0.1ml

 View more versions of this product

Product Code. 18662172

  • £366.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POLR3K Polyclonal antibody specifically detects POLR3K in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLR3K
Polyclonal
Western Blot 1:500-1:2000
C11, DNA-directed RNA polymerase III subunit K, DNA-directed RNA polymerase III subunit RPC10, DNA-directed RNA polymerases III 12.5 kDa polypeptide, hRPC11, HsC11p, polymerase (RNA) III (DNA directed) polypeptide K (12.3 kDa), polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa, RNA polymerase III 12.5 kDa subunit, RNA polymerase III subunit (hRPC11), RNA polymerase III subunit C10, RNA polymerase III subunit C11, RNA polymerase III subunit CII, RPC10, RPC11C11-RNP3, RPC12.5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-108 of human POLR3K (NP_057394.3). MLLFCPGCGNGLIVEEGQRCHRFACNTCPYVHNITRKVTNRKYPKLKEVDDVLGGAAAWENVDSTAESCPKCEHPRAYFMQLQTRSADEPMTTFYKCCNAQCGHRWRD
0.1 mL
Core ESC Like Genes, Stem Cell Markers
51728
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.