missing translation for 'onlineSavingsMsg'
Learn More

POLR3K Antibody (3F5), Novus Biologicals™

Product Code. 18349659 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349659 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349659 Supplier Novus Biologicals Supplier No. H00051728M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POLR3K Monoclonal antibody specifically detects POLR3K in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3K
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 3F5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11932
Gene Alias C11, DNA-directed RNA polymerase III subunit K, DNA-directed RNA polymerase III subunit RPC10, DNA-directed RNA polymerases III 12.5 kDa polypeptide, hRPC11, HsC11p, polymerase (RNA) III (DNA directed) polypeptide K (12.3 kDa), polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa, RNA polymerase III 12.5 kDa subunit, RNA polymerase III subunit (hRPC11), RNA polymerase III subunit C10, RNA polymerase III subunit C11, RNA polymerase III subunit CII, RPC10, RPC11C11-RNP3, RPC12.5
Host Species Mouse
Immunogen POLR3K (AAH11932, 1 a.a. ~ 108 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLLFCPGCGNGLIVEEGQRCHRFACNTCPYVHNITRKVTNRKYPKLKEVDDVLGGAAAWENVDSTAESCPKCEHPRAYFMQLQTRSADEPMTTFYKCCNAQCGHRWRD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51728
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.