missing translation for 'onlineSavingsMsg'
Learn More

POLR3GL Antibody (OTI5E8), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18417657 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417657 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417657 Supplier Novus Biologicals Supplier No. NBP273523AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POLR3GL Monoclonal antibody specifically detects POLR3GL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3GL
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI5E8
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias alternative RNA polymerase III subunit 32, DNA-directed RNA polymerase III subunit G-like, DNA-directed RNA polymerase III subunit RPC7-like, flj32422, FLJ34890, MGC3200, polymerase (RNA) III (DNA directed) polypeptide G (32kD) like, polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like, RNA polymerase III subunit C7-like, RPC32HOM, RPC32-like protein
Host Species Mouse
Immunogen Full length recombinant protein of human POLR3GL (NP_115681) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84265
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.