Learn More
POLR3G Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24701-1-AP-150UL
This item is not returnable.
View return policy
Description
The POLR3G antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human POLR3G. This antibody recognizes human antigen. The POLR3G antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| POLR3G | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10622 | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| 627 μg/mL | |
| Rabbit | |
| Primary | |
| Human |