missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR3F Polyclonal Antibody
Description
A suggested positive control is human brain tissue lysate. PA5-20589 can be used with blocking peptide PEP-0709.
The human POLR3F is a component of RNA III polymerase. RNA polymerase III transcribes many essential, small, noncoding RNAs, including the 5S rRNAs and tRNAs. While most pol III-transcribed genes are found scattered throughout the linear chromosome maps or in multiple linear clusters, there is increasing evidence that many of these genes prefer to be spatially clustered, often at or near the nucleolus. This association could create an environment that fosters the coregulation of transcription by pol III with transcription of the large ribosomal RNA repeats by RNA polymerase I (pol I) within the nucleolus. Given the high number of pol III-transcribed genes in all eukaryotic genomes, the spatial organization of these genes is likely to affect a large portion of the other genes in a genome. POLR3F has also been recently identified as an HIV dependency factor (HDF), suggesting that POLR3F may be an important drug target in HIV treatment. At least two isoforms of POLR3F are known to exist.
Specifications
Specifications
| Antigen | POLR3F |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | POLR3F |
| Gene Accession No. | Q921X6, Q9H1D9 |
| Gene Alias | 2810411G20Rik; 3010019O03Rik; 3110032A07Rik; DNA-directed RNA polymerase III subunit F; DNA-directed RNA polymerase III subunit RPC6; Polr3f; polymerase (RNA) III (DNA directed) polypeptide F; polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa; polymerase (RNA) III subunit F; RNA polymerase III 39 kDa subunit; RNA polymerase III C39 subunit; RNA polymerase III subunit C6; RNA polymerase III subunit F; RPC39; RPC6 |
| Gene Symbols | POLR3F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?