Learn More
POLR3E Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15022-1-AP-20UL
 View more versions of this product
Product Code. 16863994
  • £135.00 / 20µL
Description
The POLR3E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human POLR3E. This antibody recognizes human, mouse, rat antigen. The POLR3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
POLR3E
Polyclonal
Unconjugated
20 μL
55718
IgG
Western Blot, Immunoprecipitation, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse