missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19886
This item is not returnable.
View return policy
Description
POLR3E Polyclonal specifically detects POLR3E in Human samples. It is validated for Western Blot.
Specifications
| POLR3E | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| POLR3E | |
| Recombinant protein encompassing a sequence within the center region of human POLR3E. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55718 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| DNA-directed RNA polymerase III 80 kDa polypeptide, DNA-directed RNA polymerase III subunit RPC5, FLJ10509, KIAA1452, polymerase (RNA) III (DNA directed) polypeptide E (80kD), RNA polymerase III 80 kDa subunit RPC5, RNA polymerase III subunit C5, RPC5, SIN | |
| Rabbit | |
| 80 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction