missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR3D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575463
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a protein which interacts with the N-terminal region of BRCA1. In addition to its ability to bind BRCA1 in vivo and in vitro, it shares homology with the 2 most conserved regions of BRCA1: the N-terminal RING motif and the C-terminal BRCT domain. The RING motif is a cysteine-rich sequence found in a variety of proteins that regulate cell growth, including the products of tumor suppressor genes and dominant protooncogenes. This protein also contains 3 tandem ankyrin repeats. The BARD1/BRCA1 interaction is disrupted by tumorigenic amino acid substitutions in BRCA1, implying that the formation of a stable complex between these proteins may be an essential aspect of BRCA1 tumor suppression. This protein may be the target of oncogenic mutations in breast or ovarian cancer.
Specifications
| POLR3D | |
| Polyclonal | |
| Unconjugated | |
| Polr3d | |
| 2810426M17Rik; 44kDa; AI326118; AW489084; BN51; BN51 (BHK21) temperature sensitivity complementing; Bn51t; DNA-directed RNA polymerase III 47 kDa polypeptide; DNA-directed RNA polymerase III subunit C4; DNA-directed RNA polymerase III subunit D; DNA-directed RNA polymerase III subunit RPC4; HOX1C; Polr3d; polymerase (RNA) III (DNA directed) polypeptide D; polymerase (RNA) III (DNA directed) polypeptide D, 44kDa; polymerase (RNA) III subunit D; Protein BN51; RNA polymerase III 47 kDa subunit; RNA polymerase III subunit C4; RNA polymerase III subunit D; RPC4; RPC53; RPC53 homolog; temperature sensitive complementation, cell cycle specific, tsBN51; TSBN51 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 306012, 661, 67065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P05423, Q91WD1 | |
| Polr3d | |
| Synthetic peptide corresponding to amino acids 281-330 of Human POLR3D. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction