missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ POLR3D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29237
This item is not returnable.
View return policy
Description
POLR3D Polyclonal antibody specifically detects POLR3D in Human samples. It is validated for Immunohistochemistry
Specifications
| POLR3D | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 661 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| BN51, BN51 (BHK21) temperature sensitivity complementing, BN51TDNA-directed RNA polymerase III 47 kDa polypeptide, DNA-directed RNA polymerase III subunit D, DNA-directed RNA polymerase III subunit RPC4, polymerase (RNA) III (DNA directed) polypeptide D, 44kDa, Protein BN51, RNA polymerase III 47 kDa subunit, RNA polymerase III subunit C4, RPC4, RPC53 homolog, temperature sensitive complementation, cell cycle specific, tsBN51, TSBN51RPC53 | |
| Between 175 and 225 | |
| 10 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction