missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR3C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555640
This item is not returnable.
View return policy
Description
Immunogen sequence: QHFRDQAIVS AVANRMDQTS SEIVRTMLRM SEITTSSSAP FTQPLSSNEI FRSLPVGYNI SKQVLDQYLT LLADDPLEFV GKSG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3C is a specific core component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. POLR3C may direct with other members of the subcomplex RNA Pol III binding to the TFIIIB-DNA complex via the interactions between TFIIIB and POLR3F. POLR3C may be involved either in the recruitment and stabilization of the subcomplex within RNA polymerase III, or in stimulating catalytic functions of other subunits during initiation. POLR3C plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses.POLR3C acts as nuclear and cytosolic DNA sensor involved in innate immune response. POLR3C can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF- Kappa-B through the RIG-I pathway.
Specifications
| POLR3C | |
| Polyclonal | |
| Unconjugated | |
| POLR3C | |
| 4933407E01Rik; DNA-directed III 62 kDa polypeptide; DNA-directed RNA polymerase III subunit C; DNA-directed RNA polymerase III subunit RPC3; hypothetical protein LOC507314; POLR3C; polymerase (RNA) III (DNA directed) polypeptide C; polymerase (RNA) III (DNA directed) polypeptide C (62kD); polymerase (RNA) III subunit C; RNA polymerase III 62 kDa subunit; RNA polymerase III subunit C; RNA polymerase III subunit C3; RPC3; RPC62 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10623, 310685, 74414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XIL3, Q9BUI4, Q9D483 | |
| POLR3C | |
| Recombinant protein corresponding to Human POLR3C. Recombinant protein control fragment (Product #RP-94513). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction