missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94211-0.1ml
This item is not returnable.
View return policy
Description
POLR3A Polyclonal antibody specifically detects POLR3A in Human samples. It is validated for Western Blot
Specifications
| POLR3A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA-directed RNA polymerase III largest subunit, DNA-directed RNA polymerase III subunit A, DNA-directed RNA polymerase III subunit RPC1, EC 2.7.7.6, hRPC155, POL3A, polymerase (RNA) III (DNA directed) polypeptide A, 155kDa, RNA polymerase III 155 kDa subunit, RNA polymerase III subunit C160, RNA polymerase III subunit RPC155-D, RPC1, RPC155RNA polymerase III subunit C1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human POLR3A (NP_008986.2). MVKEQFRETDVAKKISHICFGMKSPEEMRQQAHIQVVSKNLYSQDNQHAPLLYGVLDHRMGTSEKDRPCETCGKNLADCLGHYGYIDLELPCFHVGYFRAVIGILQMICKTCCHIMLSQEEKKQFLDYLKRPGLTYLQKRGLKKKISDKCRKKNICHHCGAFNGTVKKCGLLKIIHEKYKTNKKVVDPIVSNFLQSFETA | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11128 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction