missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2K Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117366
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. POLR2K (polymerase (RNA) II (DNA directed) polypeptide K), also known as RPB12, RPABC4, RPB7.0, hRPB7.0, hsRPB10a, RPB10alpha or ABC10-alpha, is a 58 amino acid nuclear protein belonging to the archaeal rpoP/eukaryotic RPC10 RNA polymerase subunit family. The gene encoding POLR2K is located on human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies.
Specifications
| POLR2K | |
| Polyclonal | |
| Unconjugated | |
| POLR2K | |
| ABC10-alpha; DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide; DNA-directed RNA polymerase II subunit K; DNA-directed RNA polymerases I, II, and III subunit RPABC4; hRPB7.0; hsRPB10a; MafY; mCG145114-like; metallothionein-I gene transcription activator; Mt1a; POLR2K; polymerase (RNA) II (DNA directed) polypeptide K; polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa; polymerase (RNA) II subunit K; RNA polymerase II 7.0 kDa subunit; RNA polymerase II subunit K; RNA polymerases I, II, and III subunit ABC4; RPABC4; RPB10alpha; RPB12; RPB7.0 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5440 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.61 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P53803 | |
| POLR2K | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human POLR2K. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction