missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2K Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568172
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. POLR2K (polymerase (RNA) II (DNA directed) polypeptide K), also known as RPB12, RPABC4, RPB7.0, hRPB7.0, hsRPB10a, RPB10alpha or ABC10-alpha, is a 58 amino acid nuclear protein belonging to the archaeal rpoP/eukaryotic RPC10 RNA polymerase subunit family. The gene encoding POLR2K is located on human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies.
Specifications
| POLR2K | |
| Polyclonal | |
| Unconjugated | |
| POLR2K | |
| ABC10-alpha; DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide; DNA-directed RNA polymerase II subunit K; DNA-directed RNA polymerases I, II, and III subunit RPABC4; hRPB7.0; hsRPB10a; MafY; mCG145114-like; metallothionein-I gene transcription activator; Mt1a; POLR2K; polymerase (RNA) II (DNA directed) polypeptide K; polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa; polymerase (RNA) II subunit K; RNA polymerase II 7.0 kDa subunit; RNA polymerase II subunit K; RNA polymerases I, II, and III subunit ABC4; RPABC4; RPB10alpha; RPB12; RPB7.0 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17749, 5440 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.87% NaCl, 30% glycerol and 0.01% sodium azide; pH 7.3 | |
| P53803, Q63871 | |
| POLR2K | |
| Synthesized peptide derived from human POLR2K. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction