missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2J3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549326
This item is not returnable.
View return policy
Description
POLR2J3 Polyclonal Antibody for Western Blot
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.
Specifications
| POLR2J3 | |
| Polyclonal | |
| Unconjugated | |
| POLR2J3 | |
| DNA-directed RNA polymerase II subunit 11; DNA-directed RNA polymerase II subunit J2; DNA-directed RNA polymerase II subunit J3; DNA-directed RNA polymerase II subunit RPB11-b1; DNA-directed RNA polymerase II subunit RPB11-b2; POLR2J2; POLR2J3; polymerase (RNA) II (DNA directed) polypeptide J3; polymerase (RNA) II subunit J3; RNA polymerase II subunit B11-b1; RNA polymerase II subunit B11-b2; RNA polymerase II subunit J3; RPB11b1; RPB11b2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 548644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H1A7 | |
| POLR2J3 | |
| KLH conjugated synthetic peptide between 1-25 amino acids from the N-terminal region of human POLR2J3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction