missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2J2 Antibody (OTI3G4), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73520DL594
This item is not returnable.
View return policy
Description
POLR2J2 Monoclonal antibody specifically detects POLR2J2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| POLR2J2 | |
| Monoclonal | |
| DyLight 594 | |
| DNA directed RNA polymerase II polypeptide J-related, DNA-directed RNA polymerase II subunit RPB11-b1, HRPB11B, MGC105050, MGC54043, polymerase (RNA) II (DNA directed) polypeptide J2, RNA polymerase II subunit B11-b1, RPB11, RPB11b1DNA-directed RNA polymerase II subunit J2 | |
| Full length recombinant protein of human POLR2J2 (NP_116581) produced in HEK293T cells. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 246721 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3G4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction