missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2G Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-37777
This item is not returnable.
View return policy
Description
POLR2G Polyclonal specifically detects POLR2G in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| POLR2G | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DNA directed RNA polymerase II 19 kda polypeptide, DNA-directed RNA polymerase II 19 kDa polypeptide, DNA-directed RNA polymerase II subunit G, DNA-directed RNA polymerase II subunit RPB7, hRPB19, hsRPB7, MGC138369, polymerase (RNA) II (DNA directed) polypeptide G, RNA polymerase II 19 kDa subunit, RNA polymerase II seventh subunit, RNA polymerase II subunit B7, RPB19, RPB7MGC138367 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5436 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:1000 | |
| NP_002687.1 | |
| POLR2G | |
| Peptide with sequence EFDPNSNPPCYK corresponding to internal region according to NP_002687.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Sheep. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction