missing translation for 'onlineSavingsMsg'
Learn More

POLR2F Antibody (OTI2F2) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30688429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688429 Supplier Novus Biologicals Supplier No. NBP344902JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POLR2F Monoclonal antibody specifically detects POLR2F in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2F
Applications Western Blot
Classification Monoclonal
Clone OTI2F2
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias DNA-directed RNA polymerase II subunit F, DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide, HRBP14.4, II, and III subunit RPABC2, polymerase (RNA) II (DNA directed) polypeptide F, RNA Polymerase II subunit 14.4 kD, RNA polymerases I, II, and III subunit ABC2, RPB6, RPB6 homolog
Host Species Mouse
Immunogen Full length human recombiant protein of human POLR2F (NP_068809) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5435
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.