missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2F Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93621-0.1ml
This item is not returnable.
View return policy
Description
POLR2F Polyclonal antibody specifically detects POLR2F in Mouse samples. It is validated for Western Blot
Specifications
| POLR2F | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA-directed RNA polymerase II subunit F, DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide, HRBP14.4, II, and III subunit RPABC2, polymerase (RNA) II (DNA directed) polypeptide F, RNA Polymerase II subunit 14.4 kD, RNA polymerases I, II, and III subunit ABC2, RPB6, RPB6 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human POLR2F (NP_068809.1). MSDNEDNFDGDDFDDVEEDEGLDDLENAEEEGQENVEILPSGERPQANQKRITTPYMTKYERARVLGTRALQIAMCAPVMVELEGETDPLLIAMKELKARKIPIIIRRYLPDGSYEDWGVDELIITD | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 5435 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction