missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16093-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3′ untranslated region of the transcript from this gene.Specifications
| POLR2D | |
| Polyclonal | |
| Unconjugated | |
| POLR2D | |
| HSRBP4, HSRPB4, POLR2D, RNA polymerase II RPB4, RNA polymerase II subunit B4, RPB16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 364834, 5433, 69241 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15514, Q9D7M8 | |
| Polr2d | |
| POLR2D Fusion Protein Ag8701 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction